Anti-CD40 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12047-500EA 406 EUR
BIRBORB12047-500 BIRBORB12047-100
Anti-CD40 Mouse Monoclonal Antibody
Anticuerpos
FACS: 1:50-100

Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR