Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB12045-50EA 240 EUR
BIRBORB12045-50 BIRBORB12045-100
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR