Anti-CD34 Mouse Monoclonal Antibody (Biotin)

Proveedor: Biorbyt
BIRBORB12036-100EA 240 EUR
BIRBORB12036-100 BIRBORB12036-500
Anti-CD34 Mouse Monoclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR