Anti-CD34 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12032-100EA 218 EUR
BIRBORB12032-100 BIRBORB12032-500
Anti-CD34 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR