Anti-CD33 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12029-100EA 218 EUR
BIRBORB12029-100 BIRBORB12029-500
Anti-CD33 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR