Anti-FCGR2A Mouse Monoclonal Antibody (FITC (Fluorescein))
Proveedor: Biorbyt
BIRBORB12024-100EA 0 EUR
BIRBORB12024-100 BIRBORB12024-50
Anti-FCGR2A Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: FCGR2A (Fc fragment of IgG, low affinity IIa, receptor (CD32))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human