Anti-PECAM1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12020-500EA 406 EUR
BIRBORB12020-500 BIRBORB12020-100
Anti-PECAM1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR