Anti-MS4A1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB11996-500EA 406 EUR
BIRBORB11996-500 BIRBORB11996-100
Anti-MS4A1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR