Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Proveedor: Biorbyt
BIRBORB11992-50EA 269 EUR
BIRBORB11992-50 BIRBORB11992-100
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Anticuerpos
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR