Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
BIRBORB11972-100EA 327 EUR
BIRBORB11972-100 BIRBORB11972-50
Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR