Anti-ITGAL Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB11956-100EA 0 EUR
BIRBORB11956-100 BIRBORB11956-500
Anti-ITGAL Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR