Anti-CD8A Mouse Monoclonal Antibody (Biotin)
Proveedor: Biorbyt
BIRBORB11949-100EA 0 EUR
BIRBORB11949-100 BIRBORB11949-500
Anti-CD8A Mouse Monoclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human