Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB11945-100EA 327 EUR
BIRBORB11945-100 BIRBORB11945-50
Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR