Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Proveedor: Biorbyt
BIRBORB11931-100EA 0 EUR
BIRBORB11931-100 BIRBORB11931-50
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Anticuerpos
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human