Anti-TGFBR2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11667-100EA 0 EUR
BIRBORB11667-100
Anti-TGFBR2 Rabbit Polyclonal Antibody
Anticuerpos
0.1-0.5 ?g/ml

Type: Primary
Antigen: TGFBR2 (transforming growth factor, beta receptor II (70/80kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR