Anti-PTPN11 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11662-100EA 344 EUR
BIRBORB11662-100
Anti-PTPN11 Rabbit Polyclonal Antibody
Anticuerpos
0.1-0.5 ?g/ml

Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR