Anti-BRAF Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB11650-100EA 344 EUR
BIRBORB11650-100
Anti-BRAF Rabbit polyclonal antibody
Anticuerpos
0.1-0.5 ?g/ml

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR