Anti-ARRB2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11647-100EA 344 EUR
BIRBORB11647-100
Anti-ARRB2 Rabbit Polyclonal Antibody
Anticuerpos
0.1-0.5 ?g/ml

Type: Primary
Antigen: ARRB2 (arrestin, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR