Anti-UIS4 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11636-100EA 240 EUR
BIRBORB11636-100
Anti-UIS4 Goat Polyclonal Antibody
Anticuerpos
Immunofluorescence: 1:250-1:1000

Type: Primary
Antigen: UIS4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR