Anti-RAB5B Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11629-100EA 240 EUR
BIRBORB11629-100
Anti-RAB5B Goat Polyclonal Antibody
Anticuerpos
Western Blot: 1:250-1:1000

Type: Primary
Antigen: RAB5B (RAB5B, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR