Anti-RAB27A Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11622-100EA 0 EUR
BIRBORB11622-100
Anti-RAB27A Goat Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:1000; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: RAB27A (RAB27A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR