Anti-GAPDH Goat polyclonal antibody

Proveedor: Biorbyt
BIRBORB11616-100EA 240 EUR
BIRBORB11616-100
Anti-GAPDH Goat polyclonal antibody
Anticuerpos
Western Blot: 1:500-1:2000

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR