Anti-GST Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11609-100EA 0 EUR
BIRBORB11609-100
Anti-GST Goat Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000

Type: Primary
Antigen: GST
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR