Anti-CANX Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11598-100EA 240 EUR
BIRBORB11598-100
Anti-CANX Goat Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000; Immunofluorescence: 1:50-1:500

Type: Primary
Antigen: CANX (calnexin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR