Anti-TRNAP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11523-100EA 334 EUR
BIRBORB11523-100 BIRBORB11523-200
Anti-TRNAP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TRNAP1 (transfer RNA proline 1 (anticodon AGG))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR