Anti-STAT3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11431-100EA 0 EUR
BIRBORB11431-100 BIRBORB11431-200
Anti-STAT3 Rabbit Polyclonal Antibody
Anticuerpos
  • ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR