Anti-TRIM21 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB11417-100EA 312 EUR
BIRBORB11417-100 BIRBORB11417-200
Anti-TRIM21 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: TRIM21 (tripartite motif containing 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human