Anti-PSPA Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11402-100EA 312 EUR
BIRBORB11402-100 BIRBORB11402-200
Anti-PSPA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PSPA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR