Anti-RXR alpha Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11341-200EA 0 EUR
BIRBORB11341-200 BIRBORB11341-100
Anti-RXR alpha Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: RXR alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR