Anti-PTHLH Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11320-100EA 0 EUR
BIRBORB11320-100 BIRBORB11320-200
Anti-PTHLH Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PTHLH (parathyroid hormone-like hormone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR