Anti-PSAT1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB11311-5EA 51 EUR
BIRBORB11311-5 BIRBORB11311-100 BIRBORB11311-20 BIRBORB11311-200
Anti-PSAT1 Mouse Monoclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR