Anti-PDGFRB Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB11257-100EA 0 EUR
BIRBORB11257-100 BIRBORB11257-200
Anti-PDGFRB Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now