Anti-NANOG Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11097-100EA 312 EUR
BIRBORB11097-100 BIRBORB11097-5 BIRBORB11097-200 BIRBORB11097-20
Anti-NANOG Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR