Anti-Mage-1/Hla-A1 Protein Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB11016-100EA 312 EUR
BIRBORB11016-100 BIRBORB11016-200
Anti-Mage-1/Hla-A1 Protein Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Mage-1/Hla-A1 Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR