Anti-GNRH1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB10987-200EA 508 EUR
BIRBORB10987-200 BIRBORB10987-100
Anti-GNRH1 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GNRH1 (gonadotropin-releasing hormone 1 (luteinizing-releasing hormone))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR