Anti-MKI67 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB10954-200EA 522 EUR
BIRBORB10954-200 BIRBORB10954-100
Anti-MKI67 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR