Anti-IGFBP2 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB10883-200EA 0 EUR
BIRBORB10883-200 BIRBORB10883-100
Anti-IGFBP2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: IGFBP2 (insulin-like growth factor binding protein 2, 36kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat