Anti-HCV-NS1 Rabbit polyclonal antibody
Proveedor: Biorbyt
BIRBORB10788-200EA 0 EUR
BIRBORB10788-200 BIRBORB10788-100
Anti-HCV-NS1 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HCV-NS1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus