Anti-H1N1 HA2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB10765-100EA 312 EUR
BIRBORB10765-100 BIRBORB10765-200
Anti-H1N1 HA2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: H1N1 HA2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR