Anti-GRB2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB10737-20EA 109 EUR
BIRBORB10737-20 BIRBORB10737-200 BIRBORB10737-5 BIRBORB10737-100
Anti-GRB2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR