Anti-GHRH Rabbit polyclonal antibody
Proveedor: Biorbyt
BIRBORB10714-200EA 0 EUR
BIRBORB10714-200 BIRBORB10714-100
Anti-GHRH Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GHRH (growth hormone releasing hormone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat