Anti-EPHA2 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB10598-200EA 0 EUR
BIRBORB10598-200 BIRBORB10598-100
Anti-EPHA2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EPHA2 (EPH receptor A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat