Anti-DTNBP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB10567-100EA 312 EUR
BIRBORB10567-100 BIRBORB10567-200
Anti-DTNBP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DTNBP1 (dystrobrevin binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR