Anti-CD46 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB10329-200EA 0 EUR
BIRBORB10329-200 BIRBORB10329-100
Anti-CD46 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CD46 (CD46 molecule, complement regulatory protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human