Anti-CD34 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB10317-100EA 446 EUR
BIRBORB10317-100 BIRBORB10317-200
Anti-CD34 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR