Anti-PECAM1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB10314-100EA 0 EUR
BIRBORB10314-100 BIRBORB10314-200
Anti-PECAM1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 2 ?g/ml; Immunochemistry: 1:500; Immunofluorescence: 1:200

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR