Anti-Brucella Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB10209-200EA 508 EUR
BIRBORB10209-200 BIRBORB10209-100
Anti-Brucella Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: Brucella
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR