Anti-PPP1R12A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Proveedor: Biorbyt
BIRBORB9839-100EA 0 EUR
BIRBORB9839-100
Anti-PPP1R12A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: PPP1R12A (protein phosphatase 1, regulatory subunit 12A)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now