Anti-LATS1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB9151-100EA 385 EUR
BIRBORB9151-100
Anti-LATS1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: LATS1 (large tumor suppressor kinase 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR