Anti-Insulin (ID4:) Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB9023-200EA 508 EUR
BIRBORB9023-200 BIRBORB9023-100
Anti-Insulin (ID4:) Mouse Monoclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Insulin (ID4:)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR